missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Thio-D-glucose, 97+%

Manufacturer:  Alfa Aesar™ J63621MA

 View more versions of this product

Catalog No. AAJ63621MA



Specifications

Specifications

5-Thio-D-glucose
135°C to 138°C
20408-97-3
MFCD00006663
14,9334
IJJLRUSZMLMXCN-SLPGGIOYSA-N
(2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
88527
Powder
White
10mg
C6H12O5S
1865193
5-thio-d-glucose, d-glucose, 5-thio, alpha-d-glucothiopyranose, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-mercaptohexanal, unii-3prv1384uo, 5-thio-alpha-d-glucopyranose, glucose, 5-thio-, d, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
C(C(C(C(C(C=O)O)O)O)S)O
196.217
196.22
≥97%

Chemical Identifiers

20408-97-3
196.217
IJJLRUSZMLMXCN-SLPGGIOYSA-N
88527
C(C(C(C(C(C=O)O)O)O)S)O
C6H12O5S
MFCD00006663
5-thio-d-glucose, d-glucose, 5-thio, alpha-d-glucothiopyranose, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-mercaptohexanal, unii-3prv1384uo, 5-thio-alpha-d-glucopyranose, glucose, 5-thio-, d, 2r,3r,4s,5r-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
(2R,3R,4S,5R)-2,3,4,6-tetrahydroxy-5-sulfanylhexanal
Safety and Handling

Safety and Handling

RTECSNumber : LZ7500000

Documents