missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Thiazolemethanol, TCI America™

Manufacturer:  TCI America T27795G

 View more versions of this product

Catalog No. T27795G



Specifications

Specifications

5-Thiazolemethanol
Yellow
38585-74-9
MFCD04115732
WKBQQWDVVHGWDB-UHFFFAOYSA-N
1,3-thiazol-5-ylmethanol
2763216
Clear Liquid at 20°C
106°C
5g
C4H5NOS
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl thiazole, 5-hydroxymethyl-1,3-thiazole
C1=C(SC=N1)CO
115.15
115.15
≥98.0% (GC)

Chemical Identifiers

38585-74-9
115.15
WKBQQWDVVHGWDB-UHFFFAOYSA-N
2763216
C1=C(SC=N1)CO
C4H5NOS
MFCD04115732
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl thiazole, 5-hydroxymethyl-1,3-thiazole
1,3-thiazol-5-ylmethanol
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6483

TSCA : No

Recommended Storage : Freezer

Documents