Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butylpyrogallol, 97%

$105.58 - $1,176.01

Chemical Identifiers

Chemical Name or Material 5-tert-Butylpyrogallol
CAS 20481-17-8
Molecular Formula C10H14O3
Formula Weight 182.22
MDL Number MFCD00059613
Synonym 5-tert-Butyl-1,2,3-trihydroxybenzene; 1-tert-Butyl-3,4,5-trihydroxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2502206

 
alfa aesar™
B2502206
5g Each for $105.58

AAB2502214

 
alfa aesar™
B2502214
25g Each for $377.12

AAB2502222

 
alfa aesar™
B2502222
100g Each for $1,176.01
Description & Specifications

Specifications

Formula Weight 182.22
Chemical Name or Material 5-tert-Butylpyrogallol
Assay 97%
Molecular Formula C10H14O3
MDL Number MFCD00059613
CAS 20481-17-8
Synonym 5-tert-Butyl-1,2,3-trihydroxybenzene; 1-tert-Butyl-3,4,5-trihydroxybenzene
Health Hazard 1 Warning
Melting Point 133° to 136°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.