missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(tert-Butylcarbamoyl)pyridine-3-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H5308103

 View more versions of this product

Catalog No. AAH5308103



Specifications

Specifications

5-(tert-Butylcarbamoyl)pyridine-3-boronic acid
1g
C10H15BN2O3
5-tert-butylcarbamoyl pyridin-3-yl boronic acid, 5-tert-butylcarbamoyl pyridine-3-boronic acid, 3-t-butyl 5-borononicotinamide, 5-borono-n-tert-butyl nicotinamide, 5-tert-butylcarbamoyl pyridin-3-ylboronic acid, 5-tert-butylcarbamoyl pyridin-3-yl boronic acid, acmc-209rak, 3-tert-butyl 5-borononicotinamide, 5-t-butylcarbamoyl-3-pyridineboronic acid, 5-t-butylcarbamoyl pyridine-3-boronic acid
B(C1=CC(=CN=C1)C(=O)NC(C)(C)C)(O)O
222.051
222.05
245°C to 250°C
913835-99-1
MFCD08689533
FOJHBAIYLMKAIB-UHFFFAOYSA-N
[5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid
44119356
98%

Chemical Identifiers

913835-99-1
222.051
FOJHBAIYLMKAIB-UHFFFAOYSA-N
44119356
B(C1=CC(=CN=C1)C(=O)NC(C)(C)C)(O)O
C10H15BN2O3
MFCD08689533
5-tert-butylcarbamoyl pyridin-3-yl boronic acid, 5-tert-butylcarbamoyl pyridine-3-boronic acid, 3-t-butyl 5-borononicotinamide, 5-borono-n-tert-butyl nicotinamide, 5-tert-butylcarbamoyl pyridin-3-ylboronic acid, 5-tert-butylcarbamoyl pyridin-3-yl boronic acid, acmc-209rak, 3-tert-butyl 5-borononicotinamide, 5-t-butylcarbamoyl-3-pyridineboronic acid, 5-t-butylcarbamoyl pyridine-3-boronic acid
[5-(tert-butylcarbamoyl)pyridin-3-yl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents