missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5308003

 View more versions of this product

Catalog No. AAH5308003



Specifications

Specifications

5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid
1g
C11H15BFNO3
5-tert-butylcarbamoyl-2-fluorophenyl boronic acid, n-t-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorobenzeneboronic acid, n-tert-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorophenylboronic acid, 5-tert-butylcarbamoyl-2-fluorophenyl boronic acid, acmc-209qms, 5-tert-butylcarbamoyl-2-fluorophenyl boronicacid, 5-tert-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-tert-butylamino-oxomethyl-2-fluorophenyl boronic acid
B(C1=C(C=CC(=C1)C(=O)NC(C)(C)C)F)(O)O
239.053
239.05
142°C to 144°C
874289-51-7
MFCD08436054
KKSGPXWRHLBCCW-UHFFFAOYSA-N
[5-(tert-butylcarbamoyl)-2-fluorophenyl]boronic acid
44717479
97%

Chemical Identifiers

874289-51-7
239.053
KKSGPXWRHLBCCW-UHFFFAOYSA-N
44717479
B(C1=C(C=CC(=C1)C(=O)NC(C)(C)C)F)(O)O
C11H15BFNO3
MFCD08436054
5-tert-butylcarbamoyl-2-fluorophenyl boronic acid, n-t-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorobenzeneboronic acid, n-tert-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorophenylboronic acid, 5-tert-butylcarbamoyl-2-fluorophenyl boronic acid, acmc-209qms, 5-tert-butylcarbamoyl-2-fluorophenyl boronicacid, 5-tert-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-tert-butylamino-oxomethyl-2-fluorophenyl boronic acid
[5-(tert-butylcarbamoyl)-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents