Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butyl-2-methylthiophenol, 94%

Chemical Identifiers

Chemical Name or Material 5-tert-Butyl-2-methylthiophenol
CAS 7340-90-1
Molecular Formula C11H16S
Formula Weight 180.32g/mol
MDL Number MFCD00142942
UN Number UN3082
Beilstein 7473482
Synonym 5-tert-Butyl-o-toluenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1921214

 
alfa aesar™
L19212-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1921222

 
alfa aesar™
L19212-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Boiling Point 252° to 257°C
Melting Point −10°C
Flash Point 116°C (240°F)
Odor Aromatic
Quantity 25g
Chemical Name or Material 5-tert-Butyl-2-methylthiophenol
CAS 7340-90-1
Assay 94%
Molecular Formula C11H16S
Formula Weight 180.32g/mol
MDL Number MFCD00142942
UN Number UN3082
Beilstein 7473482
Synonym 5-tert-Butyl-o-toluenethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304-H361-H373-H226-H315-H319-H317-H336
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H361-H373-H226-H315-H319-H317-H336
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.