missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butyl-2-methoxybenzoic acid, 98%

$97.78 - $356.23

Chemical Identifiers

CAS 73469-54-2
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD02093964
InChI Key ZXMIDYAIOHHLRW-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methoxybenzoic acid, 2-methoxy-5-tert-butylbenzoic acid, benzoic acid, 5-1,1-dimethylethyl-2-methoxy, 5-t-butyl-2-methoxybenzoic acid, 5-tert-butyl-2-methoxybenzoicacid, 5-tert-butyl-2-methoxy-benzoic acid, 5-tert-butyl-2-methoxybenzoic acid,
PubChem CID 7010315
IUPAC Name 5-tert-butyl-2-methoxybenzoic acid
SMILES CC(C)(C)C1=CC(=C(C=C1)OC)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1783303 View Documents Alfa Aesar™
L1783303
1g Each for $97.78
 
AAL1783306 View Documents Alfa Aesar™
L1783306
5g Each for $356.23
 
Specifications

Chemical Identifiers

73469-54-2
208.257
ZXMIDYAIOHHLRW-UHFFFAOYSA-N
7010315
CC(C)(C)C1=CC(=C(C=C1)OC)C(=O)O
C12H16O3
MFCD02093964
5-tert-butyl-2-methoxybenzoic acid, 2-methoxy-5-tert-butylbenzoic acid, benzoic acid, 5-1,1-dimethylethyl-2-methoxy, 5-t-butyl-2-methoxybenzoic acid, 5-tert-butyl-2-methoxybenzoicacid, 5-tert-butyl-2-methoxy-benzoic acid, 5-tert-butyl-2-methoxybenzoic acid,
5-tert-butyl-2-methoxybenzoic acid

Specifications

74°C to 76°C
73469-54-2
MFCD02093964
5-tert-butyl-2-methoxybenzoic acid, 2-methoxy-5-tert-butylbenzoic acid, benzoic acid, 5-1,1-dimethylethyl-2-methoxy, 5-t-butyl-2-methoxybenzoic acid, 5-tert-butyl-2-methoxybenzoicacid, 5-tert-butyl-2-methoxy-benzoic acid, 5-tert-butyl-2-methoxybenzoic acid,
CC(C)(C)C1=CC(=C(C=C1)OC)C(=O)O
208.257
208.26
5-tert-Butyl-2-methoxybenzoic acid
1g
C12H16O3
2722665
ZXMIDYAIOHHLRW-UHFFFAOYSA-N
5-tert-butyl-2-methoxybenzoic acid
7010315
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning