Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butyl-2-methoxybenzeneboronic acid, 98+%

$172.41 - $172.41

Chemical Identifiers

Chemical Name or Material 5-tert-Butyl-2-methoxybenzeneboronic acid
CAS 128733-85-7
Molecular Formula C11H17BO3
Formula Weight 208.07
MDL Number MFCD06201034
Beilstein 4181890
Synonym 5-tert-Butyl-2-methoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2041503

  SDS
alfa aesar™
L2041503
1g Each for $172.41
Description & Specifications

Specifications

Formula Weight 208.07
Assay ≥98%
CAS 128733-85-7
Molecular Formula C11H17BO3
MDL Number MFCD06201034
Beilstein 4181890
Chemical Name or Material 5-tert-Butyl-2-methoxybenzeneboronic acid
Synonym 5-tert-Butyl-2-methoxyphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 103° to 106°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.