missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butyl-2-methoxyaniline, 97%

Manufacturer:  Alfa Aesar™ H5501914

 View more versions of this product

Catalog No. AAH5501914



Specifications

Specifications

5-tert-Butyl-2-methoxyaniline
113°C (235°F)
29°C to 30°C
3535-88-4
MFCD00026299
DLTLFDQLMFMTRQ-UHFFFAOYSA-N
5-tert-butyl-2-methoxyaniline
4176318
97%
0.988
204°C to 208°C
25g
C11H17NO
2-amine-4-tert-butylanisole
CC(C)(C)C1=CC(=C(C=C1)OC)N
179.263
179.26

Chemical Identifiers

3535-88-4
179.263
DLTLFDQLMFMTRQ-UHFFFAOYSA-N
4176318
CC(C)(C)C1=CC(=C(C=C1)OC)N
C11H17NO
MFCD00026299
2-amine-4-tert-butylanisole
5-tert-butyl-2-methoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents