Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-tert-Butyl-1,2,3-trimethylbenzene, 97%

$123.00 - $483.34

Chemical Identifiers

Chemical Name or Material 5-tert-Butyl-1,2,3-trimethylbenzene
CAS 98-23-7
Molecular Formula C13H20
Formula Weight 176.29
MDL Number MFCD00043642
Beilstein 2076153
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0594309

 
alfa aesar™
L0594309
10g Each for $123.00

AAL0594318

 
alfa aesar™
L0594318
50g Each for $483.34
Description & Specifications

Specifications

Formula Weight 176.29
Chemical Name or Material 5-tert-Butyl-1,2,3-trimethylbenzene
CAS 98-23-7
TSCA TSCA
Assay 97%
Molecular Formula C13H20
MDL Number MFCD00043642
Boiling Point 114° to 115°C (17mmHg)
Melting Point 31° to 33°C
Quantity 10g
Beilstein 2076153