missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Sulfosalicylic Acid Dihydrate 98.0+%, TCI America™

Manufacturer:  TCI America S083025G

 View more versions of this product

Catalog No. S083025G



Specifications

Specifications

5-Sulfosalicylic Acid Dihydrate [for Biochemical Research]
113°C
5965-83-3
MFCD00149540
BHDKTFQBRFWJKR-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid;dihydrate
2723734
Crystal-Powder at 20°C
White
25g
C7H10O8S
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O.O.O
254.209
218.18
≥98.0% (HPLC,T)

Chemical Identifiers

5965-83-3
254.209
BHDKTFQBRFWJKR-UHFFFAOYSA-N
2723734
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O.O.O
C7H10O8S
MFCD00149540
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
2-hydroxy-5-sulfobenzoic acid;dihydrate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2114

RTECSNumber : VO6500000

TSCA : Yes

Documents