missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(5-Pyrrolidin-1-ylpyrid-2-yl)methanol, 97%, Maybridge

Chemical Identifiers

CAS 930110-98-8
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H14N2O
Formula Weight 178.23
MDL Number MFCD09702371
Chemical Name or Material (5-Pyrrolidin-1-ylpyrid-2-yl)methanol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC52909DA SDS Fisher Scientific Company
CC52909DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC52909DE SDS Fisher Scientific Company
CC52909DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

930110-98-8
100.0
C10H14N2O
MFCD09702371
95%
94.5
95%
178.23
(5-Pyrrolidin-1-ylpyrid-2-yl)methanol

Specifications

1g
94.5
Amber Glass Bottle
C10H14N2O
MFCD09702371
95%
930110-98-8
100.0
95%
178.23
(5-Pyrrolidin-1-ylpyrid-2-yl)methanol

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning