Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Pyrimidinylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material 5-Pyrimidinylzinc bromide
Formula Weight 224.37
MDL Number MFCD11915900
UN Number UN3399
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58089AD

 
alfa aesar™
H58089AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.37
Chemical Name or Material 5-Pyrimidinylzinc bromide
Concentration 0.5M in THF
Sensitivity Air Sensitive
Physical Form Liquid
MDL Number MFCD11915900
UN Number UN3399
Health Hazard 1 Danger
Quantity 50mL
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Color Yellow to brown to black
Packaging ChemSeal™ bottles

Solubility
Reacts with water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with oxidizing agents, acid chlorides. Protect from humidity and water. Do not together with acids.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C