missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Pyrimidinylboronic acid, 97%, may contain varying amounts of anhydride, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451700250
Specifications
5-Pyrimidinylboronic acid | |
>300.0°C | |
109299-78-7 | |
C4H5BN2O2 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
pyrimidin-5-ylboronic acid | |
2795193 | |
97% |
Glass bottle | |
25g | |
96% min. (HPLC) | |
5-pyrimidine boronic acid, 5-pyrimidineboronic acid, 5-pyrimidinylboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-boronic acid, pyrimidin-5-yl boronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidine-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidinyl-5-boronic acid | |
B(C1=CN=CN=C1)(O)O | |
123.91 | |
123.91 |
Chemical Identifiers
109299-78-7 | |
123.91 | |
5-pyrimidine boronic acid, 5-pyrimidineboronic acid, 5-pyrimidinylboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-boronic acid, pyrimidin-5-yl boronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidine-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidinyl-5-boronic acid | |
pyrimidin-5-ylboronic acid |
C4H5BN2O2 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
2795193 | |
B(C1=CN=CN=C1)(O)O |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS Signal Word: Warning