missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Pyrimidinylboronic acid, 97%, may contain varying amounts of anhydride, Thermo Scientific™
Supplier: Thermo Fisher Scientific 451701000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-Pyrimidinylboronic acid | |
>300.0°C | |
C4H5BN2O2 | |
Glass bottle | |
5-pyrimidine boronic acid, 5-pyrimidineboronic acid, 5-pyrimidinylboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-boronic acid, pyrimidin-5-yl boronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidine-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidinyl-5-boronic acid | |
OB(O)C1=CN=CN=C1 | |
123.91 | |
123.91 |
109299-78-7 | |
96% min. (HPLC) | |
MFCD03002366,MFCD16661236 | |
100g | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
pyrimidin-5-ylboronic acid | |
2795193 | |
97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS Signal Word: Warning
RUO – Research Use Only