missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Pyrazolone, TCI America™

Manufacturer:  TCI America P25631G

 View more versions of this product

Catalog No. P25631G



Specifications

Specifications

5-Pyrazolone
≥98.0% (GC)
MFCD00127223
White-Yellow
2,4-Dihydro-3H-pyrazol-3-one, 1H-Pyrazol-5(4H)-one
C1C=NNC1=O
84.078
84.08
137-44-0
C3H4N2O
162°C
1g
ZRHUHDUEXWHZMA-UHFFFAOYSA-N
1,4-dihydropyrazol-5-one
316289
Crystal-Powder at 20°C

Chemical Identifiers

137-44-0
84.078
ZRHUHDUEXWHZMA-UHFFFAOYSA-N
316289
C1C=NNC1=O
C3H4N2O
MFCD00127223
2,4-Dihydro-3H-pyrazol-3-one, 1H-Pyrazol-5(4H)-one
1,4-dihydropyrazol-5-one
Safety and Handling

Safety and Handling

TSCA : No

Documents