Please login to your online account to display your discounted pricing

5-Piperidinothiophene-2-carbaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC72604CB

 View more versions of this product

Catalog No. CC72604CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 195.28
Quantity 250mg
Assay 97%
CAS Max % 100.0
Molecular Formula C10H13NOS
Chemical Name or Material 5-Piperidinothiophene-2-carbaldehyde
MDL Number MFCD05263177
CAS Min % 96.5
Packaging Amber Glass Bottle
CAS 24372-48-3
Assay Percent Range 97%

Specifications

Chemical Name or Material 5-Piperidinothiophene-2-carbaldehyde
CAS 24372-48-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H13NOS
Formula Weight 195.28
MDL Number MFCD05263177