missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Phenyltetrazole, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC263720250

 View more versions of this product

Catalog No. AC263720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Phenyltetrazole
Glass bottle
25g
98.5
99%
MFCD00022388
Solubility in water: .. Other solubilities: soluble in organic solvents
C1=CC=C(C=C1)C2=NNN=N2
146.15
146.15
99%
Authentic
White
18039-42-4
100.0
C7H6N4
1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, 5-phenyl tetrazole, 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 5-phenyltetrazole van, expandex 5pt, expandex ox 5pt, kempore 50xpt, phenyltetrazole
MARUHZGHZWCEQU-UHFFFAOYSA-N
5-phenyl-2H-tetrazole
87425
Crystalline Powder

Chemical Identifiers

18039-42-4
146.15
MARUHZGHZWCEQU-UHFFFAOYSA-N
87425
C1=CC=C(C=C1)C2=NNN=N2
C7H6N4
MFCD00022388
1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, 5-phenyl tetrazole, 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 5-phenyltetrazole van, expandex 5pt, expandex ox 5pt, kempore 50xpt, phenyltetrazole
5-phenyl-2H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Risk of explosion if heated under confinement.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-950-8

SDS Documents