Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride, tech. 90%

Chemical Identifiers

Chemical Name or Material 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride
CAS 166964-37-0
Purity Grade Technical Grade
Molecular Formula C10H7ClO4S3
Formula Weight 322.81
MDL Number MFCD00052143
UN Number UN3261
Beilstein 7818655
Synonym 5-(Benzenesulfonyl)thiophene-2-sulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1171303

  SDS
alfa aesar™
L1171303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL11713MD

  SDS
alfa aesar™
L11713MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 322.81
Chemical Name or Material 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00052143
CAS 166964-37-0
Synonym 5-(Benzenesulfonyl)thiophene-2-sulfonyl chloride
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 102° to 105°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3261
Molecular Formula C10H7ClO4S3
Health Hazard 1 Danger
Beilstein 7818655
Purity Grade Technical Grade
Quantity 1g
Assay 90%
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.