missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride, tech. 90%

Manufacturer:  Alfa Aesar™ L11713MD

 View more versions of this product

Catalog No. AAL11713MD



Specifications

Specifications

5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride
250mg
C10H7ClO4S3
UN3261
5-phenylsulfonyl thiophene-2-sulfonyl chloride, 5-phenylsulfonyl thiophene-2-sulfonylchloride, 5-benzenesulfonyl thiophene-2-sulfonyl chloride, 2-benzenesulfonylthiophene-5-sulfonyl chloride, 5-phenylthiophene-2,5-disulphonyl chloride, pubchem5455, 5-benzenesulphonylthiophene-2-sulphonyl chloride, acmc-1c42s, 5-phenylsulphonyl thiophene-2-sulphonyl chloride, 2-phenylsulfonylthiophene-5-sulfonyl chloride
C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(S2)S(=O)(=O)Cl
322.792
322.81
Technical
102°C to 105°C
166964-37-0
MFCD00052143
7818655
GSLVRDAYMHUGTF-UHFFFAOYSA-N
5-(benzenesulfonyl)thiophene-2-sulfonyl chloride
580877
90%

Chemical Identifiers

166964-37-0
322.792
GSLVRDAYMHUGTF-UHFFFAOYSA-N
580877
C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(S2)S(=O)(=O)Cl
C10H7ClO4S3
MFCD00052143
5-phenylsulfonyl thiophene-2-sulfonyl chloride, 5-phenylsulfonyl thiophene-2-sulfonylchloride, 5-benzenesulfonyl thiophene-2-sulfonyl chloride, 2-benzenesulfonylthiophene-5-sulfonyl chloride, 5-phenylthiophene-2,5-disulphonyl chloride, pubchem5455, 5-benzenesulphonylthiophene-2-sulphonyl chloride, acmc-1c42s, 5-phenylsulphonyl thiophene-2-sulphonyl chloride, 2-phenylsulfonylthiophene-5-sulfonyl chloride
5-(benzenesulfonyl)thiophene-2-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Documents