Please login to your online account to display your discounted pricing

5-Phenyl-o-anisidine, 98+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC198270010

 View more versions of this product

Catalog No. AC198270010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Coarse Powder
Assay ≥98.0%
Formula Weight 199.25
Quantity 1g
Color Brown
EINECS Number 254-639-7
Melting Point 81° to 84°C
Beilstein 13, III, 1946
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 5-Phenyl-o-anisidine
Packaging Glass bottle
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C13H13NO
Linear Formula C6H5C6H3(OCH3)NH2
MDL Number MFCD00007790
CAS 39811-17-1
Synonym 2-Methoxy-5-phenylaniline
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Phenyl-o-anisidine
Name Note 98+%
CAS 39811-17-1
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C13H13NO
Linear Formula C6H5C6H3(OCH3)NH2
Formula Weight 199.25
MDL Number MFCD00007790
Synonym 2-Methoxy-5-phenylaniline
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Suspected of causing cancer.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents