missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Phenyl-5H-pyrido[3,2-b]indole, TCI America™

Manufacturer:  TCI America P2578200MG

 View more versions of this product

Catalog No. P2578200MG



Specifications

Specifications

5-Phenyl-5H-pyrido[3,2-b]indole
101°C
1541200-53-6
9-Phenyl-delta-carboline
Crystal-Powder at 20°C
White-Yellow
200mg
C17H12N2
244.30
≥98.0% (GC)

Chemical Identifiers

1541200-53-6
C17H12N2
9-Phenyl-delta-carboline
Safety and Handling

Safety and Handling

TSCA : No

Documents