missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Phenyl-5H-pyrido[3,2-b]indole, TCI America™

Manufacturer:  TCI America P25781G

 View more versions of this product

Catalog No. P25781G



Specifications

Specifications

5-Phenyl-5H-pyrido[3,2-b]indole
101°C
1541200-53-6
C17H12N2
244.30
White-Yellow
1g
≥98.0% (GC)
9-Phenyl-delta-carboline
Crystal-Powder at 20°C

Chemical Identifiers

1541200-53-6
C17H12N2
9-Phenyl-delta-carboline
Safety and Handling

Safety and Handling

TSCA : No

Documents