missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Phenyl-2-thiophenesulfonyl chloride, ≥95%, Maybridge

Chemical Identifiers

CAS 97272-02-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H7ClO2S2
Formula Weight 258.75
MDL Number MFCD06797460
Chemical Name or Material 5-Phenyl-2-thiophenesulfonyl chloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC10503CB SDS Fisher Scientific Company
CC10503CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC10503DA SDS Fisher Scientific Company
CC10503DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97272-02-1
100.0
C10H7ClO2S2
MFCD06797460
95%
94.5
95%
258.75
5-Phenyl-2-thiophenesulfonyl chloride

Specifications

258.75
100.0
C10H7ClO2S2
94.5
Amber Glass Bottle
95%
250mg
95%
MFCD06797460
97272-02-1
5-Phenyl-2-thiophenesulfonyl chloride

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger