missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Phenyl-2-thiophenecarboxylic Acid, TCI America™

Manufacturer:  TCI America P23471G

 View more versions of this product

Catalog No. P23471G



Specifications

Specifications

5-Phenyl-2-thiophenecarboxylic Acid
190°C
19163-24-7
MFCD01313432
QGMFBCDNJUZQBZ-UHFFFAOYSA-N
5-phenylthiophene-2-carboxylic acid
736494
Crystal-Powder at 20°C
White-Yellow
1g
C11H8O2S
5-phenyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-phenyl, 5-phenyl-2-thiophene carboxylic acid, pubchem19533, 5-phenyl-2-thenoic acid, 2-carboxy-5-phenylthiophene, 5-phenyl 2-thiophenecarboxylic acid, 5-phenyl-thiophene-2-carboxylic acid
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)O
204.243
204.24
≥98.0% (GC,T)

Chemical Identifiers

19163-24-7
204.243
QGMFBCDNJUZQBZ-UHFFFAOYSA-N
736494
C1=CC=C(C=C1)C2=CC=C(S2)C(=O)O
C11H8O2S
MFCD01313432
5-phenyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-phenyl, 5-phenyl-2-thiophene carboxylic acid, pubchem19533, 5-phenyl-2-thenoic acid, 2-carboxy-5-phenylthiophene, 5-phenyl 2-thiophenecarboxylic acid, 5-phenyl-thiophene-2-carboxylic acid
5-phenylthiophene-2-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents