missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Phenyl-2-furoyl chloride

Chemical Identifiers

CAS 57489-93-7
Molecular Formula C11H7ClO2
Formula Weight 206.63
MDL Number MFCD02258033
UN Number UN3261
Synonym 5-Phenylfuran-2-carbonyl chloride
Chemical Name or Material 5-Phenyl-2-furoyl chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5040303 SDS Alfa Aesar™
H5040303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5040306 SDS Alfa Aesar™
H5040306
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57489-93-7
206.63
UN3261
5-Phenyl-2-furoyl chloride
C11H7ClO2
MFCD02258033
5-Phenylfuran-2-carbonyl chloride

Specifications

1g
C11H7ClO2
MFCD02258033
5-Phenylfuran-2-carbonyl chloride
57489-93-7
206.63
UN3261
5-Phenyl-2-furoyl chloride

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.