missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Phenyl-1H-tetrazole, 99%

$61.19 - $144.96

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7H6N4
Formula Weight 146.15
MDL Number MFCD00022388
Chemical Name or Material 5-Phenyl-1H-tetrazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2566414 SDS Alfa Aesar™
B2566414
25g Each for $61.19
AAB2566422 SDS Alfa Aesar™
B2566422
100g Each for $144.96
Specifications Description & Specifications

Chemical Identifiers

18039-42-4
146.15
5-Phenyl-1H-tetrazole
C7H6N4
MFCD00022388

Specifications

Odorless
25g
C7H6N4
MFCD00022388
99%
≈215°C (dec.)
18039-42-4
146.15
5-Phenyl-1H-tetrazole

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : XF7538750

TSCA : TSCA