Learn More
5-Phenyl-1H-tetrazole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2566414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Phenyl-1H-tetrazole | |
∼215°C (decomposition) | |
C7H6N4 | |
25 g | |
MARUHZGHZWCEQU-UHFFFAOYSA-N | |
5-phenyl-2H-tetrazole | |
87425 | |
99% |
18039-42-4 | |
Odorless | |
MFCD00022388 | |
5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van | |
C1=CC=C(C=C1)C2=NNN=N2 | |
146.153 | |
146.15 |
Chemical Identifiers
18039-42-4 | |
146.153 | |
MARUHZGHZWCEQU-UHFFFAOYSA-N | |
87425 | |
C1=CC=C(C=C1)C2=NNN=N2 |
C7H6N4 | |
MFCD00022388 | |
5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van | |
5-phenyl-2H-tetrazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P264b-P270-P280-P301+P312-P330-P374-P501c
H228-H302
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 241-950-8
RTECSNumber : XF7538750
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only