Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Phenyl-1-pentenylboronic acid pinacol ester, 96%

$102.05 - $283.23

Chemical Identifiers

Chemical Name or Material 5-Phenyl-1-pentenylboronic acid pinacol ester
CAS 154820-97-0
Molecular Formula C17H25BO2
Formula Weight 272.2
MDL Number MFCD03788752
Beilstein 7295229
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1970103

  SDS
alfa aesar™
L1970103
1g Each for $283.23

AAL19701MD

  SDS
alfa aesar™
L19701MD
250mg Each for $102.05
Description & Specifications

Specifications

Formula Weight 272.2
Chemical Name or Material 5-Phenyl-1-pentenylboronic acid pinacol ester
Assay 96%
Molecular Formula C17H25BO2
MDL Number MFCD03788752
CAS 154820-97-0
Beilstein 7295229
Quantity 1g

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.