Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Phenyl-1,3,4-oxadiazole-2-thiol, 98%

$112.51 - $1,397.66

Chemical Identifiers

Chemical Name or Material 5-Phenyl-1,3,4-oxadiazole-2-thiol
CAS 3004-42-0
Molecular Formula C8H6N2OS
Formula Weight 178.21
MDL Number MFCD00030467
Synonym 2-Mercapto-5-phenyl-1,3,4-oxadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1661814

 
alfa aesar™
A1661814
25g Each for $112.51

AAA1661822

 
alfa aesar™
A1661822
100g Each for $330.95

AAA1661836

 
alfa aesar™
A1661836
500g Each for $1,397.66
Description & Specifications

Specifications

Formula Weight 178.21
Chemical Name or Material 5-Phenyl-1,3,4-oxadiazole-2-thiol
TSCA TSCA
Assay 98%
Molecular Formula C8H6N2OS
MDL Number MFCD00030467
CAS 3004-42-0
Synonym 2-Mercapto-5-phenyl-1,3,4-oxadiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 220°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RO0829900
Quantity 25g

Applications
5-Phenyl-1,3,4-oxadiazole-2-thiol is used to produce 1-Benzoyl-4-(4-chlor-phenyl)-thiosemicarbazid.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.