Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Phenyl-1,3,4-oxadiazole-2-methylamine hydrochloride, 96%

$84.99 - $345.05

Chemical Identifiers

Chemical Name or Material 5-Phenyl-1,3,4-oxadiazole-2-methylamine hydrochloride
CAS 1187931-09-4
Molecular Formula C9H9N3O·HCl
Formula Weight 211.6
MDL Number MFCD09028173
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5103206

  SDS
alfa aesar™
H5103206
5g Each for $345.05

AAH5103203

  SDS
alfa aesar™
H5103203
1g Each for $84.99
Description & Specifications

Specifications

Formula Weight 211.6
Chemical Name or Material 5-Phenyl-1,3,4-oxadiazole-2-methylamine hydrochloride
Health Hazard 1 Warning
Quantity 5g
CAS 1187931-09-4
MDL Number MFCD09028173
Molecular Formula C9H9N3O·HCl
Assay 96%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.