missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5 Percent Ruthenium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
$105.12 - $1536.38
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
18-607-976
|
Spectrum Chemical Mfg Cor
SP4015G |
5 g |
Each for $105.12
|
|
|||||
18-607-977
|
Spectrum Chemical Mfg Cor
SP40125G |
25 g |
Each for $426.88
|
|
|||||
18-607-978
|
Spectrum Chemical Mfg Cor
SP401100G |
100 g |
Each for $1,536.38
|
|
|||||
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
[Ru] |
Ru | |
MFCD00011207 MFCD03458417 | |
ruthenium |
Specifications
7440-18-8 | |
Ru | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium |
5 g | |
MFCD00011207 MFCD03458417 | |
[Ru] | |
101.07 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.