missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5 Percent Palladium on Calcium Carbonate Powder, Unreduced, Dry, 3 microns, BASF Catalyst, Spectrum™ Chemical
$167.50 - $2384.67
Chemical Identifiers
| CAS | 5-3-7440 |
|---|---|
| Molecular Formula | Pd |
| Molecular Weight (g/mol) | 106.42 |
| MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
| InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
| IUPAC Name | palladium |
| SMILES | [Pd] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-607-945
|
Spectrum Chemical Mfg Cor
SP1095G |
5 g |
Each for $167.50
|
|
|||||
|
18-607-946
|
Spectrum Chemical Mfg Cor
SP10925G |
25 g |
Each for $672.43
|
|
|||||
|
18-607-947
|
Spectrum Chemical Mfg Cor
SP109100G |
100 g |
Each for $2,384.67
|
|
|||||
Chemical Identifiers
| 5-3-7440 | |
| 106.42 | |
| KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
| [Pd] |
| Pd | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| palladium |
Specifications
| 5-3-7440 | |
| Pd | |
| KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
| palladium |
| 5 g | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| [Pd] | |
| 106.42 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.