missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(p-Tolyl)-1H-tetrazole, TCI America™

Manufacturer:  TCI America T248625G

 View more versions of this product

Catalog No. T248625G



Specifications

Specifications

5-(p-Tolyl)-1H-tetrazole
248°C
24994-04-5
MFCD01318164
BCCJIAZPYBJASR-UHFFFAOYSA-N
5-(4-methylphenyl)-2H-tetrazole
285164
Crystal-Powder at 20°C
White-Yellow
25g
C8H8N4
5-4-methylphenyl-1h-tetrazole, 5-p-tolyl-1h-tetrazole, 5-4-methylphenyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetrazole, 5-p-tolyl-2h-tetrazole, 5-p-tolyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetraazole, 5-p-methylphenyl-1h-tetrazole, 5-p-tolyl-1h-tetrazole, 5-4-methylphenyl tetrazole
CC1=CC=C(C=C1)C2=NNN=N2
160.18
160.18
≥98.0% (HPLC,T)

Chemical Identifiers

24994-04-5
160.18
BCCJIAZPYBJASR-UHFFFAOYSA-N
285164
CC1=CC=C(C=C1)C2=NNN=N2
C8H8N4
MFCD01318164
5-4-methylphenyl-1h-tetrazole, 5-p-tolyl-1h-tetrazole, 5-4-methylphenyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetrazole, 5-p-tolyl-2h-tetrazole, 5-p-tolyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetraazole, 5-p-methylphenyl-1h-tetrazole, 5-p-tolyl-1h-tetrazole, 5-4-methylphenyl tetrazole
5-(4-methylphenyl)-2H-tetrazole
Safety and Handling

Safety and Handling

TSCA : No

Documents