Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Oxo-5-(2-thienyl)valeric acid, 97%

$46.64 - $138.54

Chemical Identifiers

Chemical Name or Material 5-Oxo-5-(2-thienyl)valeric acid
CAS 22971-62-6
Molecular Formula C9H10O3S
Formula Weight 198.24
MDL Number MFCD00151848
Beilstein 141775
Synonym 5-Oxo-5-(2-thienyl)pentanoic acid; 4-(2-Thenoyl)butyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1336206

 
alfa aesar™
L1336206
5g Each for $138.54

AAL1336203

 
alfa aesar™
L1336203
1g Each for $46.64
Description & Specifications

Specifications

Formula Weight 198.24
Chemical Name or Material 5-Oxo-5-(2-thienyl)valeric acid
Assay 97%
Molecular Formula C9H10O3S
MDL Number MFCD00151848
CAS 22971-62-6
Beilstein 141775
Synonym 5-Oxo-5-(2-thienyl)pentanoic acid; 4-(2-Thenoyl)butyric acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Melting Point 94° to 97°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.