missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Oxo-5-(2-thienyl)valeric acid, 97%

Manufacturer:  Alfa Aesar™ L1336206

 View more versions of this product

Catalog No. AAL1336206



Specifications

Specifications

5-Oxo-5-(2-thienyl)valeric acid
5g
C9H10O3S
141775
GSQMRVFOHNXKOR-UHFFFAOYSA-N
5-oxo-5-thiophen-2-ylpentanoic acid
255155
97%
94°C to 97°C
22971-62-6
MFCD00151848
5-oxo-5-2-thienyl valeric acid, 5-oxo-5-thiophen-2-yl pentanoic acid, 4-2-thienoyl butyric acid, 5-oxo-5-thien-2-ylpentanoic acid, 5-oxo-5-2-thienyl pentanoic acid, 2-thiophenepentanoicacid, d-oxo, acmc-1cj8s, 4-2-thenoyl butyric acid, 5-2-thienoyl butyric acid, 5-thien-2-yl-5-oxopentanoic acid
C1=CSC(=C1)C(=O)CCCC(=O)O
198.236
198.24

Chemical Identifiers

22971-62-6
198.236
GSQMRVFOHNXKOR-UHFFFAOYSA-N
255155
C1=CSC(=C1)C(=O)CCCC(=O)O
C9H10O3S
MFCD00151848
5-oxo-5-2-thienyl valeric acid, 5-oxo-5-thiophen-2-yl pentanoic acid, 4-2-thienoyl butyric acid, 5-oxo-5-thien-2-ylpentanoic acid, 5-oxo-5-2-thienyl pentanoic acid, 2-thiophenepentanoicacid, d-oxo, acmc-1cj8s, 4-2-thenoyl butyric acid, 5-2-thienoyl butyric acid, 5-thien-2-yl-5-oxopentanoic acid
5-oxo-5-thiophen-2-ylpentanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents