Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose

$63.11 - $160.08

Chemical Identifiers

Chemical Name or Material 5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose
CAS 68703-51-5
Molecular Formula C14H28O5Si
Formula Weight 304.46
MDL Number MFCD01075716
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1598603

 
alfa aesar™
L1598603
1g Each for $160.08

AAL15986MD

 
alfa aesar™
L15986MD
250mg Each for $63.11
Description & Specifications

Specifications

Formula Weight 304.46
Chemical Name or Material 5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose
Molecular Formula C14H28O5Si
MDL Number MFCD01075716
CAS 68703-51-5
Quantity 1g
Optical Rotation −13.3° (c=1 in Chloroform)
Recommended Storage 2° to 8°C
Melting Point 50° to 53°C

Applications
5-O-tert-Butyldimethylsilyl-2,3-O-isopropylidene-α,β-D-ribofuranose is used as a starting material for the preparation of N-benzyl alfa-L-manno aminocyclopentitol , 4-deoxy-N-benzyl-alfa-L-manno aminocyclopentitol and N-benzyl 6,7-di-epi-trehalamine using stereoselective allylation. It is also used in oligosaccharides.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Incompatible with oxidizing agents.

Safety and Handling

2° to 8°C