missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%

Manufacturer:  Alfa Aesar™ 4481509

 View more versions of this product

Catalog No. AA4481509



Specifications

Specifications

5'-O- (4,4'-Dimethoxytrityl)thymidine
40615-39-2
MFCD00010113
UBTJZUKVKGZHAD-UPRLRBBYSA-N
1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-5-methylpyrimidine-2,4-dione
162419
Powder
10g
C31H32N2O7
5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
CC1=CN(C(=O)NC1=O)C2CC(C(O2)COC(C3=CC=CC=C3)(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)O
544.604
544.6
≥98%

Chemical Identifiers

40615-39-2
544.604
UBTJZUKVKGZHAD-UPRLRBBYSA-N
162419
CC1=CN(C(=O)NC1=O)C2CC(C(O2)COC(C3=CC=CC=C3)(C4=CC=C(C=C4)OC)C5=CC=C(C=C5)OC)O
C31H32N2O7
MFCD00010113
5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-5-methylpyrimidine-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents