missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Norbornene-2-endo,3-endo-dimethanol, 98%

Manufacturer:  Alfa Aesar™ H5345406

 View more versions of this product

Catalog No. AAH5345406



Specifications

Specifications

5-Norbornene-2-endo,3-endo-dimethanol
82°C to 86°C
699-97-8
MFCD00674044
IGHHPVIMEQGKNE-WZENYGAOSA-N
[(2R,3S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
12253651
98%
170°C to 175°C (25mmHg)
5g
C9H14O2
2s,3r-3-hydroxymethyl bicyclo 2.2.1 hept-5-en-2-yl methanol
C1C2C=CC1C(C2CO)CO
154.209
154.21

Chemical Identifiers

699-97-8
154.209
IGHHPVIMEQGKNE-WZENYGAOSA-N
12253651
C1C2C=CC1C(C2CO)CO
C9H14O2
MFCD00674044
2s,3r-3-hydroxymethyl bicyclo 2.2.1 hept-5-en-2-yl methanol
[(2R,3S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents