missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Norbornen-2-methanol (mixture of isomers), TCI America™

$67.22 - $220.00

Chemical Identifiers

CAS 95-12-5
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
MDL Number MFCD00167571
InChI Key LUMNWCHHXDUKFI-UHFFFAOYSA-N
Synonym 5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, bicyclo 2.2.1 hept-5-ene-2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
PubChem CID 78946
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanol
SMILES C1C2CC(C1C=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09795G View Documents TCI America
N09795G
5g Each for $67.22
 
N097925G View Documents TCI America
N097925G
25g Each for $220.00
 
Specifications

Chemical Identifiers

95-12-5
124.183
LUMNWCHHXDUKFI-UHFFFAOYSA-N
78946
C1C2CC(C1C=C2)CO
C8H12O
MFCD00167571
5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, bicyclo 2.2.1 hept-5-ene-2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
5-bicyclo[2.2.1]hept-2-enylmethanol

Specifications

120°C
5g
C8H12O
5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, bicyclo 2.2.1 hept-5-ene-2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
C1C2CC(C1C=C2)CO
124.183
124.18
≥98.0% (GC)
Yellow
95-12-5
MFCD00167571
LUMNWCHHXDUKFI-UHFFFAOYSA-N
5-bicyclo[2.2.1]hept-2-enylmethanol
78946
Clear Liquid at 20°C
5-Norbornen-2-methanol (mixture of isomers)
Safety and Handling

Safety and Handling

TSCA : Yes