Please login to your online account to display your discounted pricing

5-Nonyloxytryptamine oxalate, Tocris


Selective 5-HT1B agonist

Quantity 10mg

Manufacturer: r & d systems  090110

 View more versions of this product

Catalog No. 090110

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 5-Nonyloxytryptamine oxalate
Quantity 10mg
CAS 157798-13-5
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C19H30N2O·C2H2O4
Formula Weight 392.49

Specifications

Chemical Name or Material 5-Nonyloxytryptamine oxalate
Assay Percent Range >99%
Molecular Formula C19H30N2O·C2H2O4
Formula Weight 392.49
Accessories

Store at Room Temperature