Please login to your online account to display your discounted pricing

5-Nonyloxytryptamine oxalate, Tocris

Selective 5-HT1B agonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material 5-Nonyloxytryptamine oxalate
Assay Percent Range >99%
Molecular Formula C19H30N2O·C2H2O4
Formula Weight 392.49
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

090110

 
r & d systems
090110
10mg Each for $201.36

090150

 
r & d systems
090150
50mg Each for $844.73
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 392.49
Molecular Formula C19H30N2O·C2H2O4
CAS 157798-13-5
Chemical Name or Material 5-Nonyloxytryptamine oxalate
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature