Please login to your online account to display your discounted pricing

5-Nonyloxytryptamine oxalate, Tocris


Selective 5-HT1B agonist

Quantity 50mg

Manufacturer: r & d systems  090150

 View more versions of this product

Catalog No. 090150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 392.49
Chemical Name or Material 5-Nonyloxytryptamine oxalate
CAS 157798-13-5
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Molecular Formula C19H30N2O·C2H2O4

Specifications

Chemical Name or Material 5-Nonyloxytryptamine oxalate
Assay Percent Range >99%
Molecular Formula C19H30N2O·C2H2O4
Formula Weight 392.49
Accessories

Store at Room Temperature