missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nonanone 98%, ACROS Organics™

Manufacturer:  Acros Organics 159761000

 View more versions of this product

Catalog No. AC159761000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Nonanone
65°C
Glass bottle
186.0°C to 187.0°C
-50.0°C
98%
97.5
98%
CH3(CH2)3CO(CH2)3CH3
01, I, 365
Solubility in water: insoluble. Other solubilities: soluble in ethanol;, very soluble in diethyl ether, chloroform.
CCCCC(=O)CCCC
142.242
142.24
98%
0.8200g/mL
Authentic
1.4180 to 1.4200
Colorless to Yellow
100mL
502-56-7
100.0
C9H18O
MFCD00009484
5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
WSGCRAOTEDLMFQ-UHFFFAOYSA-N
nonan-5-one
10405
Liquid

Chemical Identifiers

502-56-7
142.242
WSGCRAOTEDLMFQ-UHFFFAOYSA-N
10405
CCCCC(=O)CCCC
C9H18O
MFCD00009484
5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
nonan-5-one
Safety and Handling

Safety and Handling

EINECSNumber : 207-946-5

RTECSNumber : RA8230000

TSCA : TSCA

SDS Documents