Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nonanol, 98%

$44.32 - $44.32

Chemical Identifiers

Chemical Name or Material 5-Nonanol
CAS 623-93-8
Molecular Formula C9H20O
Formula Weight 144.26
MDL Number MFCD00021944
Beilstein 1733552
Synonym Di-n-butyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0581214

 
alfa aesar™
L0581214
25g Each for $44.32
Description & Specifications

Specifications

Formula Weight 144.26
Chemical Name or Material 5-Nonanol
Assay 98%
CAS 623-93-8
Molecular Formula C9H20O
MDL Number MFCD00021944
Beilstein 1733552
Synonym Di-n-butyl carbinol
Flash Point 96°C (204°F)
Boiling Point 194° to 196°C
Melting Point 5°C
Quantity 25g

Applications
5-Nonanol is used as intermediates in chemical research. It is used as natural substances and extractives.

Solubility
Soluble in water (0.46 mg/ml at 15°C), and chloroform.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.