missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitrouracil, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 226581000

 View more versions of this product

Catalog No. AC226581000



Description

Description

Specifications

Specifications

5-Nitrouracil
Glass bottle
>300.0°C
p.a.
99.0
99+%
MFCD00006021
2,4 1h,3h-pyrimidinedione, 5-nitro, 2,4 1h,3h-pyrimidinedione-5-nitro, 2,4-dihydroxy-5-nitropyrimidine, 4-hydroxy-5-nitropyrimidin-2 1h-one, 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-nitropyrimidine-2,4 1h,3h-dione, 5-nitropyrimidine-2,4-diol, 5-nitrouracil, 5-nitrouracil 2,4-dihydroxy-5-nitropyrimidine, uracil, 5-nitro
C1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
157.08
CHEBI:60763
Crystalline Powder or Needles
Authentic
White to Yellow
100g
611-08-5
100.0
C4H3N3O4
24, 320
TUARVSWVPPVUGS-UHFFFAOYSA-N
5-nitro-1H-pyrimidine-2,4-dione
69135
157.08
99+%

Chemical Identifiers

611-08-5
157.08
TUARVSWVPPVUGS-UHFFFAOYSA-N
69135
5-nitro-1H-pyrimidine-2,4-dione
C4H3N3O4
MFCD00006021
2,4 1h,3h-pyrimidinedione, 5-nitro, 2,4 1h,3h-pyrimidinedione-5-nitro, 2,4-dihydroxy-5-nitropyrimidine, 4-hydroxy-5-nitropyrimidin-2 1h-one, 5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 5-nitropyrimidine-2,4 1h,3h-dione, 5-nitropyrimidine-2,4-diol, 5-nitrouracil, 5-nitrouracil 2,4-dihydroxy-5-nitropyrimidine, uracil, 5-nitro
CHEBI:60763
C1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-250-4

TSCA : TSCA

SDS Documents