missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitrouracil, 98+%

Manufacturer:  Alfa Aesar™ A1244814

 View more versions of this product

Catalog No. AAA1244814



Specifications

Specifications

5-Nitrouracil
~300°C
25g
MFCD00006021
TUARVSWVPPVUGS-UHFFFAOYSA-N
5-nitro-1H-pyrimidine-2,4-dione
69135
157.09
611-08-5
≥98%
C4H3N3O4
10410
C1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
157.085
CHEBI:60763

Chemical Identifiers

611-08-5
157.085
TUARVSWVPPVUGS-UHFFFAOYSA-N
CHEBI:60763
C1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
C4H3N3O4
MFCD00006021
69135
5-nitro-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

RTECSNumber : UV9104545

TSCA : TSCA

SDS Documents