Please login to your online account to display your discounted pricing

5-Nitrothiophene-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Nitrothiophene-3-carboxylic acid
CAS 40357-96-8
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H3NO4S
Formula Weight 173.15
MDL Number MFCD00052582
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

DFP00046CB

  SDS
fisher scientific company
DFP00046CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H3NO4S
MDL Number MFCD00052582
CAS 40357-96-8
CAS Min % 96.5
Chemical Name or Material 5-Nitrothiophene-3-carboxylic acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle