missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitrothiophene-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0149314

 View more versions of this product

Catalog No. AAL0149314



Description

Description

5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrothiophene-2-carboxaldehyde is used as a chemical for protoemics research as well as the synthesis of 2,3-dihydro-2-(5-nitro-2-thienyl) quinazolin-4-(1H)-ones1 and various novel oxime ether derivatives which are anti-protozoan agents.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

5-Nitrothiophene-2-carboxaldehyde
72°C to 76°C
MFCD00005433
120540
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene
CHTSWZNXEKOLPM-UHFFFAOYSA-N
5-nitrothiophene-2-carbaldehyde
78281
98%
4521-33-9
C5H3NO3S
25 g
Air sensitive
Insoluble in water.
C1=C(SC(=C1)[N+](=O)[O-])C=O
157.143
157.15

Chemical Identifiers

4521-33-9
157.143
CHTSWZNXEKOLPM-UHFFFAOYSA-N
78281
C1=C(SC(=C1)[N+](=O)[O-])C=O
C5H3NO3S
MFCD00005433
5-nitrothiophene-2-carboxaldehyde, 5-nitro-2-thiophenecarboxaldehyde, 5-nitro-2-thiophenecarbaldehyde, 5-nitro-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 5-nitro, 5-nitrothiophene-2-aldehyde, thiophenecarboxaldehyde, 5-nitro, pubchem15668, 5-nitrothiophen aldehyde, 5-nitro-2-formyl-thiophene
5-nitrothiophene-2-carbaldehyde
Safety and Handling

Safety and Handling

EINECSNumber : 224-850-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only