Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitroisoquinoline, 98+%

$59.41 - $180.09

Chemical Identifiers

Chemical Name or Material 5-Nitroisoquinoline
CAS 607-32-9
Molecular Formula C9H6N2O2
Formula Weight 174.16
MDL Number MFCD00006905
Beilstein 134858
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1531106

 
alfa aesar™
A1531106
5g Each for $59.41

AAA1531114

 
alfa aesar™
A1531114
25g Each for $180.09
Description & Specifications

Specifications

Formula Weight 174.16
Chemical Name or Material 5-Nitroisoquinoline
Assay ≥98%
Molecular Formula C9H6N2O2
MDL Number MFCD00006905
CAS 607-32-9
Beilstein 134858
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 106° to 110°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.